Learn More
Sino Biological Recombinant Mouse NOTCH1 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 50654M02S50
Encompass_Preferred
A DNA sequence encoding the mouse Notch1 (NP_032740.3) (Met1-Gln526) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2274