Learn More
Sino Biological Recombinant Mouse NOTCH1 Protein (His Tag)
Supplier:  Sino Biological 50654M08B50
Encompass_Preferred
A DNA sequence encoding the mouse NOTCH1 (NP_032740.3) N-terminal fragment (Met 1-Gln 526) was expressed,with a C-terminal polyhistidine tag.
Catalog No. 50-162-2276