Learn More
Sino Biological Recombinant Mouse IFNA14 Protein (His Tag)
Supplier:  Sino Biological 50669M08H100
Encompass_Preferred
A DNA sequence encoding the mouse IFNA14 (NP_996858.1)(Cys24-Lys189) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2289