Learn More
Sino Biological Recombinant Mouse CD300A/LMIR1 Protein (hFc Tag)
Supplier:  Sino Biological 50675M02H100
Encompass_Preferred
A DNA sequence encoding the mouse CD300A (Q6SJQ0-1) (Met 1-Arg 183) extracellular domain was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2308