Learn More
Sino Biological Recombinant Mouse Phospholipase A2 IIE/PLA2G2E Protein (His Tag)
Supplier:  Sino Biological 50687M08H100
Encompass_Preferred
A DNA sequence encoding the mouse PLA2G2E (Q9QUL3) (Met 1-Cys 142) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2320