Learn More
Sino Biological Recombinant Mouse Noggin/NOG Protein (hFc Tag)
Supplier:  Sino Biological 50688M02H100
Encompass_Preferred
A DNA sequence encoding the mouse NOG (P97466) (Met1-Cys232) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2323