Learn More
Sino Biological Recombinant Mouse CD83 Protein (hFc Tag)
Supplier:  Sino Biological 50694M02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CD83 (O88324) (Met 1-Arg 133) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2338