Learn More
Sino Biological Recombinant Mouse CHODL Protein (hFc Tag)
Supplier:  Sino Biological 50699M02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CHODL (EDK98299.1) (Met 1-Asn 216) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2348