Learn More
Sino Biological Recombinant Mouse CD302 Protein (His Tag)
Supplier:  Sino Biological 50706M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CD302 isoform 2 (Q9DCG2-2) (Met 1-His 156) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2362