Learn More
Sino Biological Recombinant Mouse CLEC10A/CD301 Protein (hFc Tag)
Supplier:  Sino Biological 50707M01H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CLEC10A (NP_001191181.1) (Gln 58-Ser 305) was fused with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-2365