Learn More
Sino Biological Recombinant Mouse IFN-beta Protein (hFc Tag)
Supplier:  Sino Biological 50708M02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse IFNB1 (P01575) (Met 1-Asn 182) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2368