Learn More
Sino Biological Recombinant Mouse IFN-beta Protein
Supplier:  Sino Biological 50708MCCH100
Encompass_Preferred
A DNA sequence encoding the mouse IFNB1 (P01575) (Met 1-Asn 182) was fused with five amino acids (DDDDK) at the C-terminus was expressed and purified.
Catalog No. 50-162-2371