Learn More
Sino Biological Recombinant Mouse Siglec-3/CD33 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50712M08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CD33 (Q63994-1) (Met 1-Glu 240) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2389