Learn More
Sino Biological Recombinant Mouse Siglec-3/CD33 Protein
Supplier:  Sino Biological 50712MCCH200
Encompass_Preferred
A DNA sequence encoding the mouse Cd33 (NP_067268.1) (Met1-Glu240) was expressed with six amino acids (LEVLFQ) at the C- terminus.
Catalog No. 50-162-2390