Learn More
Sino Biological Recombinant Mouse CD24 Protein (hFc Tag)
Supplier:  Sino Biological 50717M02H100
Encompass_Preferred
A DNA sequence encoding the mouse CD24 (P24807) (Met1-Arg52) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2404