Learn More
Sino Biological Recombinant Mouse CD52 Protein (hFc Tag)
Supplier:  Sino Biological 50721M02H100
Encompass_Preferred
A DNA sequence encoding the mouse Cd52 (NP_038734.1) (Met1-Lys46) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2412