Learn More
Sino Biological Recombinant Mouse CD53 Protein (aa 107-181, His Tag)
Supplier:  Sino Biological 50722M07H100
Encompass_Preferred
A DNA sequence encoding the mouse CD53 (Q61451) (Glu107-Asn181) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-2414