Learn More
Sino Biological Recombinant Mouse Neuroligin 1 Protein (His Tag)
Supplier:  Sino Biological 50725M08H100
Encompass_Preferred
A DNA sequence encoding the mouse NLGN1 isoform 1 (Q99K10-1) extracellular domain (Met 1-Ser 697) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2420