Learn More
Sino Biological Recombinant Mouse CD69 Protein (ECD, His Tag)
Supplier:  Sino Biological 50731M08H100
Encompass_Preferred
A DNA sequence encoding the mouse Cd69 (NP_001028294.1) (Asn62-Arg199) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-2436