Learn More
Sino Biological Recombinant Mouse CD43 Protein (hFc Tag)
Supplier:  Sino Biological 50735M02H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse SPN (P15702) (Met 1-Gly 248) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2441