Learn More
Sino Biological Recombinant Mouse P-Selectin/CD62P Protein (hFc Tag)
Supplier:  Sino Biological 50737M02H100
Encompass_Preferred
A DNA sequence encoding the mouse SELP (NP_035477.1) (Met1-Ala709) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2448