Learn More
Sino Biological Recombinant Mouse ST6GAL1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50740M07H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse ST6GAL1 (NP_666045.1) (Lys 27-Cys 403) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-2452