Learn More
Sino Biological Recombinant Mouse CD81 Protein (His Tag)
Supplier:  Sino Biological 50743M07H50
Encompass_Preferred
A DNA sequence encoding the mouse Cd81 (NP_598416.1) (Lys106-Lys211) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-2459