Learn More
Sino Biological Recombinant Mouse Lactoferrin/LTF Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50750M08H50
Encompass_Preferred
A DNA sequence encoding the mouse LTF (AAH06904.1) (Met 1-Gln 707) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2463