Learn More
Sino Biological Recombinant Mouse N-Cadherin Protein
Supplier:  Sino Biological 50752MCCH100
Encompass_Preferred
A DNA sequence encoding the mouse CDH2 (P15116) (Met1-Ala724) was fused with six amino acids (LEVLFQ) at the C-terminus was expressed and purified.
Catalog No. 50-162-2464