Learn More
Sino Biological Recombinant Mouse Carbonic Anhydrase 8/CA8 Protein (His Tag)
Supplier:  Sino Biological 50767M08E100
Encompass_Preferred
A DNA sequence encoding the mouse CA8 (P28651) (Met 1-Gln 291) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2481