Learn More
Sino Biological Recombinant Mouse Carbonic Anhydrase X Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50768M08H100
Encompass_Preferred
A DNA sequence encoding the mouse Car10 (P61215) (Met 1-Asn 300) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-162-2483