Learn More
Sino Biological Recombinant Mouse PSGL-1/CD162 Protein (hFc Tag)
Supplier:  Sino Biological 50770M02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse SELPLG (Q62170) (Met 1-Cys 307) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2488