Learn More
Sino Biological Recombinant Mouse Frizzled-5 Protein (hFc Tag)
Supplier:  Sino Biological 50772M02S100
Encompass_Preferred
A DNA sequence encoding the mouse FZD5 (Q9EQD0) (Met1-Pro167) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2492