Learn More
Sino Biological Recombinant Mouse Carboxypeptidase A2 Protein (His Tag)
Supplier:  Sino Biological 50778M08H100
Encompass_Preferred
A DNA sequence encoding the mouse CPA2 (Q504N0) (Met 1-Tyr 417) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2498