Learn More
Sino Biological Recombinant Mouse DOPA Decarboxylase/DDC Protein (His Tag)
Supplier:  Sino Biological 50799M08B100
Encompass_Preferred
A DNA sequence encoding the mouse DDC (O88533) (Met 1-Glu 480) was expressed,with a C-terminal polyhistidine tag.
Catalog No. 50-162-2509