Learn More
Sino Biological Recombinant Mouse PD-L2/B7-DC Protein (hFc Tag)
Catalog No. 501622515
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse PDCD1LG2 (Q9WUL5) (Met 1-Arg 219) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2515 Supplier Sino Biological Supplier No. 50804M02H100
A DNA sequence encoding the extracellular domain of mouse PDCD1LG2 (Q9WUL5) (Met 1-Arg 219) was fused with the Fc region of human IgG1 at the C-terminus.