Learn More
Sino Biological Recombinant Mouse Klrb1a Protein (His Tag)
Supplier:  Sino Biological 50806M07H100
Encompass_Preferred
A DNA sequence encoding the secreted form of mouse Klrb1a (P27811) (Gln 67-His 227) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-2521