Learn More
Sino Biological Recombinant Mouse IL1R1 Protein (hFc Tag)
Supplier:  Sino Biological 50807M02H100
Encompass_Preferred
A DNA sequence encoding the mouse IL1R1 (P13504) extracellular domain (Met 1-Lys 338) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2523