Learn More
Sino Biological Recombinant Mouse OX40/TNFRSF4 Protein (hFc Tag)
Supplier:  Sino Biological 50808M02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse TNFRSF4 (NP_035789.1) (Met 1-Pro 211) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2527