Learn More
Sino Biological Recombinant Mouse OX40/TNFRSF4 Protein
Supplier:  Sino Biological 50808MCCH200
Encompass_Preferred
A DNA sequence encoding the mouse Tnfrsf4 (NP_035789.1) (Met1-Pro211) was expressed with six amino acids (LEVLFQ) at the C-terminus.
Catalog No. 50-162-2531