Learn More
Sino Biological Recombinant Mouse 4-1BB/CD137 Protein (soluble form, rFc Tag)
Supplier:  Sino Biological 50811M31H100
Encompass_Preferred
A DNA sequence encoding the mouse Tnfrsf9 (Met1-Leu211) was expressed with the Fc region of rabbit IgG at the C-terminus.
Catalog No. 50-162-2541