Learn More
New England Biolabs, Inc. HphI – 1000 units
SDP
Supplier:  New England Biolabs, Inc. R0158S
Encompass
A restriction endonuclease that recognizes the sequence GGTGANNNNNNN_N^.
Catalog No. 50-812-060