Learn More
New England Biolabs, Inc. EciI – 100 units
SDP
Supplier:  New England Biolabs, Inc. R0590S
Encompass
A restriction endonuclease that recognizes the sequence GGCGGANNNNNNNNN_NN^.
Catalog No. 50-812-302