Learn More
Sino Biological Recombinant Mouse SIRP-beta/Sirpb1a Protein (His Tag)
Supplier:  Sino Biological 50816M08H50
Encompass_Preferred
A DNA sequence encoding the mouse SIRPB1A (BAD26610.1) (Met 1-Lys 363) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2548