Learn More
Sino Biological Recombinant Mouse Transferrin Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 50817M02H100
Encompass_Preferred
A DNA sequence encoding the mouse TRF (Q921I1) (Met1-His697) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-2549