Learn More
Sino Biological Recombinant Mouse Transferrin Protein (His Tag)
Supplier:  Sino Biological 50817M08H100
Encompass_Preferred
A DNA sequence encoding the mouse TRF (Q921I1) (Met1-His697) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2551