Learn More
Sino Biological Recombinant Mouse DDR1 Protein (hFc Tag)
Supplier:  Sino Biological 50829M02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse DDR1 (NP_766550.1) (Met 1-Thr 414) was fused with the Fc region of human IgG1 at the C-terminus
Catalog No. 50-162-2567