Learn More
Sino Biological Recombinant Mouse DDR1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50829M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse DDR1 (NP_766550.1) (Met 1-Thr 414) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2569