Learn More
Sino Biological Recombinant Mouse CAMKIV Protein (His & GST Tag)
Supplier:  Sino Biological 50838M20B50
Encompass_Preferred
A DNA sequence encoding the mouse CAMK4 (P08414) (Met1-Tyr469) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-162-2576