Learn More
Sino Biological Recombinant Mouse IL-10RB/IL-10R beta Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50846M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse IL10RB (NP_032375.2) (Met 1-Ser 222) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2583