Learn More
Sino Biological Recombinant Mouse GLIPR1 Protein (His Tag)
Supplier:  Sino Biological 50869M08H100
Encompass_Preferred
A DNA sequence encoding the mouse GLIPR1 (NP_082884.1) (Met1-Thr223) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-2589