Learn More
Sino Biological Recombinant Mouse EPCR Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50874M08H50
Encompass_Preferred
A DNA sequence encoding the mouse PROCR (Q64695) extracellular domain (Met 1-Ser 214) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-162-2594