Learn More
Sino Biological Recombinant Mouse NEK3 Protein (His & GST Tag)
Supplier:  Sino Biological 50880M20B50
Encompass_Preferred
A DNA sequence encoding the mouse NEK3 (ABK42288.1) (Met 1-Ala 509) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-162-2598