Learn More
Sino Biological Recombinant Mouse CSK Protein (His & GST Tag)
Supplier:  Sino Biological 50893M20B50
Encompass_Preferred
A DNA sequence encoding the mouse CSK (P41241?) (Met 1-Leu 450) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-162-2601